Learn More
FGFR-2 Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 50010574
Description
Positive control: Liver carcinoma Cellular location: Cytoplasmic, membrane.
FGFR-2 Polyclonal antibody specifically detects FGFR-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| FGFR-2 | |
| Polyclonal | |
| Unconjugated | |
| FGFR2 | |
| AU043015; AW556123; bacteria-expressed kinase; BBDS; BEK; BEK fibroblast growth factor receptor; BFR-1; CD332; CEK3; CFD1; ECT1; FGF Receptor 2; FGF-10; FGFR2; FGFR-2; Fgfr7; Fgfr-7; fibroblast growth factor receptor 2; JWS; Keratinocyte growth factor receptor; KGFR; KGFRTr; KSAM; K-SAM; protein tyrosine kinase, receptor like 14; Soluble KGFR; svs; TK14; TK25 | |
| A synthetic peptide from aa 362-374 of human FGFR-2 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with proprietary stabilizer and 0.01% sodium azide | |
| P21802 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2263 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.