Learn More
FGFR-3 Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 50010234

Description
Positive control: Breast ca and umbilical cord Cellular location: Cell membrane.
FGFR-3 Polyclonal antibody specifically detects FGFR-3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western BlotSpecifications
FGFR-3 | |
Polyclonal | |
Unconjugated | |
FGFR3 | |
ACH; CD333; CEK2; FGFR3; FGFR-3; FGFR3deltaTM; Fibroblast growth factor receptor 3; fibroblast growth factor receptor 3 variant 4; Flg-2; FR3; HBGFR; heparin-binding growth factor receptor; HSFGFR3EX; hydroxyaryl-protein kinase; JTK4; Mfr3; sam3; tyrosine kinase JTK4 | |
A 14aa synthetic peptide derived from aa 359-372 of human FGFR-3 protein | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P22607 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2261 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.