Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR-4, Rabbit anti-Human, Clone: 29.2, Millipore Sigma™

Catalog No. MABC1127100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1127100 100 μL
MABC112725 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1127100 Supplier MilliporeSigma Supplier No. MABC1127100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-FGFR-4, clone 29.2, Cat. No. MABC1127, is a rabbit monoclonal antibody that detects Fibroblast growth factor receptor 4 and has been tested for use in Flow Cytometry.

Fibroblast growth factor receptor 4 (UniProt: P22455; also known as EC: 2.7.10.1, FGFR-4, CD334) is encoded by the FGRF4 (also known as JTK2, TKF) gene (Gene ID: 2264) in human. FGFR-4 is a highly conserved, single-pass type I membrane protein with tyrosine kinase activity that serves as a cell surface receptor for fibroblast growth factors and regulates cell proliferation differentiation, and migration. FGFR-4 is expressed in gastrointestinal epithelial cells, pancreas, and gastric and pancreatic cancer cell lines. In the absence of a ligand, it is present as a monomer in an inactive conformation. Upon ligand binding it homodimerizes into an active conformation with autophosphorylation of tyrosine residues. FGFR-4 is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off in the mature form. It contains an extracellular domain (aa 22-369), a transmembrane domain (aa 370-390), and a cytoplasmic domain (aa 391-802). FGFR-4 undergoes N-glycosylation, and when not fully glycosylated it can undergo proteasomal degradation. FGFR-4 signaling is down-regulated by receptor internalization and degradation. MMP14 is reported to promote internalization and degradation of FGFR-4. Mutations that lead to constitutive kinase activation or impair normal FGFR-4 inactivation lead to aberrant signaling leading to tumor growth. (Ref.: Turkington, RC et al. (2014). Cell Death Dis. 5; e1046).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFR-4
Applications Flow Cytometry
Classification Monoclonal
Clone 29.2
Conjugate Unconjugated
Dilution Flow Cytometry Analysis: A 1:4 dilution from a representative lot detected FGFR-4 in one million LNCaP cells.
Formulation Rabbit monoclonal antibody in supernatant without preservatives.
Gene Alias Fibroblast growth factor receptor 4;EC: 2.7.10.1;CD334
Gene Symbols FGFR4
Host Species Rabbit
Immunogen His-tagged recombinant extracellular domain of human fibroblast growth factor receptor 4 (FGFR-4).
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001278909
Test Specificity This rabbit monoclonal antibody detects human fibroblast growth factor receptor 4 (FGFR-4). It targets an epitope with in the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.