Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR1 alpha Antibody (1058809), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB11336MFV610
Description
FGFR1 alpha Monoclonal antibody specifically detects FGFR1 alpha in Human samples. It is validated for Flow CytometrySpecifications
FGFR1 alpha | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Human embryonic kidney cell, HEK293-derived human FGFR1 alpha, Arg22-Glu376, Accession # P11362.3 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1058809 | |
Flow Cytometry | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2260 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction