Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR1 Antibody (001), Alexa Fluor™ 532

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290469AF532
Description
FGFR1 Monoclonal specifically detects FGFR1 in Mouse samples. It is validated for ELISA.Specifications
FGFR1 | |
Monoclonal | |
Alexa Fluor 532 | |
basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, FLT-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FGFR1 (P16092-1; Met 1-Glu 376). | |
0.1 mL | |
Angiogenesis, Apoptosis, Core ESC Like Genes, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
2260 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction