Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR1 Antibody (M19B2), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001287PE
Description
FGFR1 Monoclonal specifically detects FGFR1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Product Image.Specifications
FGFR1 | |
Monoclonal | |
PE | |
P11362 | |
FGFR1 | |
Recombinant human ectodomain of FGFR1b expressed in E. coli beginning with proline 23; antigen contained NH2-terminal gly-ser-pro-gly-ile and COOH-terminal glu-phe sequences. | |
0.1 mL | |
Angiogenesis, Apoptosis, Core ESC Like Genes, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
2260 | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
M19B2 | |
Flow Cytometry | |
basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, FLT-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Reacts with both alpha and beta isoforms. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction