Learn More
Promega FGFR1 Kinase Enzyme System, 1mg
Catalog No. PRV2992 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2992 1 mg
1 options
Catalog No. PRV2992 Supplier Promega Supplier No. V2992
Add to Cart
Add to Cart
Request a Quote
Recombinant human FGFR1 (amino acids 399-822) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FGFR1 (also known as FLT2) is a member of the Fibroblast Growth Factor Receptor family.
  • Easily Screen and Profile FGFR1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C