Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FGFR1 Monoclonal Antibody (M2F12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA126256
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FGFR1 (also known as FLT2) is a member of the Fibroblast Growth Factor Receptor family that constitute a family of four membrane-spanning tyrosine kinases (FGFR1-4) which serve as high-affinity receptors for 17 growth factors (FGF1-17). The FGF Receptor family plays an important role in multiple biological processes, including mesoderm induction and patterning, cell growth and migration, organ formation and bone growth. FGFR1 is alternatively spliced generating multiple splice variants that are differentially expressed during embryo development and in the adult body. Other defects in FGFR1 are responsible for several diseases which include Pfeiffer syndrome (PS), idiopathic hypogonadotropic hypogonadism (IHH), Kallmann syndrome type 2 (KAL2), osteoglophonic dysplasia (OGD), non-syndromic trigonocephaly, Jackson-Weiss syndrome, Antley-Bixler syndrome. Chromosomal aberrations involving this gene are associated with stem cell myeloproliferative disorder and stem cell leukemia lymphoma syndrome.
Specifications
FGFR1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P11362, P16092, Q04589 | |
FGFR1 | |
Recombinant human ectodomain of FGFr1a,expressed in E. coli beginning with pro23; antigen contained NH2-terminal gly-ser-pro-gly-ile and COOH-termina. | |
125 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
M2F12 | |
Unconjugated | |
FGFR1 | |
2.7.10.1; AW208770; Basic fibroblast growth factor receptor 1; basic fibroblast growth factor receptor I; BFGFR; bFGF-R; bFGF-R-1; CD331; CD331 antigen; CEK; CEK1; cek1 protein; cek1 protein precursor; c-fgr; Eask; ECCL; FGF receptor; FGF receptor-1; FGFBR; FGFR; fgfr 1; Fgfr1; FGFR-1; FGFR1/PLAG1 fusion; FGFR1IIIC; FGFRI; Fibroblast growth factor receptor 1; fibroblast growth factor receptor 1 (fms-related tyrosine kinase 2, Pfeiffer syndrome); fibroblast growth factor receptor 1-IIIc; fibroblast growth factor receptor type 1; Flg; FLT2; FLT-2; FMS-like tyrosine kinase 2; fms-related tyrosine kinase 2; HBGFR; heparin-binding growth factor receptor; HH2; HRTFDS; Hspy; hydroxyaryl-protein kinase; KAL2; LOW QUALITY PROTEIN: fibroblast growth factor receptor 1; MFR; N-sam; OGD; proto-oncogene c-Fgr; Tyrosine kinase receptor CEK1 | |
Mouse | |
Protein G | |
RUO | |
14182, 2260, 79114 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction