Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR1 Mouse anti-Human, Mouse, Clone: 10F12E7, Proteintech
SDP

Catalog No. 501736414 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6414 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6414 Supplier Proteintech Group Inc Supplier No. 603251IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FGFR1 (fibroblast growth factor receptor 1) is a member of the fibroblast growth factor receptor family containing an Ig-like domain and a tyrosine kinase domain. FGFR1 has multiple isoforms and is a Type I membrane protein. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein consists of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of FGFR1 interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. FGFR1 binds both acidic and basic fibroblast growth factors and is involved in limb induction. FGFR1 can be modified by phosphorylation and can bind basic/acidic fibroblast factor depending on the receptor isoform. FGFR1 has been shown to interact with N-cadherin and NCAM. Four genes encoding for high affinity cell surface FGF receptors (FGFRs) have been identified: FGFR-1, FGFR-2, FGFR-3 and FGFR-4.FGFRs are members of the tyrosine kinase family of growth factor receptors. At the mRNA level, FGFR1 is highly expressed in developing human tissues including the brain, vascular basement membrane and skin. Defects in FGFR1 result in Pfeiffer syndrome associated with craniosynostosis. Other defects in FGFR1 are responsible for several diseases which include Pfeiffer syndrome (PS), idiopathic hypogonadotropic hypogonadism (IHH), Kallmann syndrome type 2 (KAL2), osteoglophonic dysplasia (OGD), non-syndromic trigonocephaly, Jackson-Weiss syndrome, Antley-Bixler syndrome. Chromosomal aberrations involving this gene are associated with stem cell myeloproliferative disorder and stem cell leukemia lymphoma syndrome.

Specifications

Antigen FGFR1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 10F12E7
Concentration 1.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FGFR1
Gene Accession No. P11362, P16092
Gene Alias bFGF R 1, BFGFR, CD331, CEK, FGFBR, FGFR 1, FGFR1, FLG, FLT 2, FLT2, Fms like tyrosine kinase 2, HBGFR, KAL2, N SAM, OGD, Proto oncogene c Fgr
Gene Symbols FGFR1
Host Species Mouse
Immunogen FGFR1 Fusion Protein Ag3646
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14182, 2260
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.