Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGFR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142830
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142830 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142830 Supplier Invitrogen™ Supplier No. PA5142830
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize reported isoforms 1, 2, 10, 11 and 14. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

FGFR1 (also known as FLT2) is a member of the Fibroblast Growth Factor Receptor family that constitute a family of four membrane-spanning tyrosine kinases (FGFR1-4) which serve as high-affinity receptors for 17 growth factors (FGF1-17). The FGF Receptor family plays an important role in multiple biological processes, including mesoderm induction and patterning, cell growth and migration, organ formation and bone growth. FGFR1 is alternatively spliced generating multiple splice variants that are differentially expressed during embryo development and in the adult body. Other defects in FGFR1 are responsible for several diseases which include Pfeiffer syndrome (PS), idiopathic hypogonadotropic hypogonadism (IHH), Kallmann syndrome type 2 (KAL2), osteoglophonic dysplasia (OGD), non-syndromic trigonocephaly, Jackson-Weiss syndrome, Antley-Bixler syndrome. Chromosomal aberrations involving this gene are associated with stem cell myeloproliferative disorder and stem cell leukemia lymphoma syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFR1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene FGFR1
Gene Accession No. P11362
Gene Alias 2.7.10.1; AW208770; Basic fibroblast growth factor receptor 1; basic fibroblast growth factor receptor I; BFGFR; bFGF-R; bFGF-R-1; CD331; CD331 antigen; CEK; CEK1; cek1 protein; cek1 protein precursor; c-fgr; Eask; ECCL; FGF receptor; FGF receptor-1; FGFBR; FGFR; fgfr 1; Fgfr1; FGFR-1; FGFR1/PLAG1 fusion; FGFR1IIIC; FGFRI; Fibroblast growth factor receptor 1; fibroblast growth factor receptor 1 (fms-related tyrosine kinase 2, Pfeiffer syndrome); fibroblast growth factor receptor 1-IIIc; fibroblast growth factor receptor type 1; Flg; FLT2; FLT-2; FMS-like tyrosine kinase 2; fms-related tyrosine kinase 2; HBGFR; heparin-binding growth factor receptor; HH2; HRTFDS; Hspy; hydroxyaryl-protein kinase; KAL2; LOW QUALITY PROTEIN: fibroblast growth factor receptor 1; MFR; N-sam; OGD; proto-oncogene c-Fgr; Tyrosine kinase receptor CEK1
Gene Symbols FGFR1
Host Species Goat
Immunogen Peptide with sequence RCRLRDDVQSIN.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2260
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.