Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR1OP Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300287AF594
Description
FGFR1OP Polyclonal specifically detects FGFR1OP in Human samples. It is validated for ELISA.Specifications
FGFR1OP | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 594 | |
FGFR1 oncogene partner, FOPfibroblast growth factor receptor 1 oncogene partner | |
Produced in rabbits immunized with purified, recombinant FGFR1OP (O95684; Ala2-Ala379). | |
0.1 mL | |
Angiogenesis, Apoptosis, Cell Cycle and Replication | |
11116 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction