Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGFR1OP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549739 Supplier Invitrogen™ Supplier No. PA549739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total FGFR1OP protein.

This gene encodes a largely hydrophilic protein postulated to be a leucine-rich protein family member. A t(6;8)(q27;p11) chromosomal translocation, fusing this gene and the fibroblast growth factor receptor 1 (FGFR1) gene, has been found in cases of myeloproliferative disorder. The resulting chimeric protein contains the N-terminal leucine-rich region of this encoded protein fused to the catalytic domain of FGFR1. This gene is thought to play an important role in normal proliferation and differentiation of the erythroid lineage. Alternatively spliced transcript variants that encode different proteins have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFR1OP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FGFR1OP
Gene Accession No. O95684, Q4V7C1, Q66JX5
Gene Alias 4930553O10Rik; centrosomal protein 43; centrosomal protein 43; FGFR1 oncogene partner; CEP43; FGFR1 oncogene partner; FGFR1 oncogene partner 1; FGFR1 oncoprotein partner-like protein; Fgfr1op; fibroblast growth factor receptor 1 oncogene partner; FOP; LOC683722; Unknown (protein for MGC:134294)
Gene Symbols FGFR1OP
Host Species Rabbit
Immunogen Synthesized peptide derived from human FGFR1 Oncogene Partner.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11116, 683722, 75296
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.