Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGFR1OP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567212
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567212 Supplier Invitrogen™ Supplier No. PA567212
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDSFFDDPIPK PEKTYGLRKE PRKQAGSLAS LSDAPPLKSG LSSLAGAPSL KDSESKRGNT VL Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

This gene encodes a largely hydrophilic protein postulated to be a leucine-rich protein family member. A t(6;8)(q27;p11) chromosomal translocation, fusing this gene and the fibroblast growth factor receptor 1 (FGFR1) gene, has been found in cases of myeloproliferative disorder. The resulting chimeric protein contains the N-terminal leucine-rich region of this encoded protein fused to the catalytic domain of FGFR1. This gene is thought to play an important role in normal proliferation and differentiation of the erythroid lineage. Alternatively spliced transcript variants that encode different proteins have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFR1OP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FGFR1OP
Gene Accession No. O95684
Gene Alias 4930553O10Rik; centrosomal protein 43; centrosomal protein 43; FGFR1 oncogene partner; CEP43; FGFR1 oncogene partner; FGFR1 oncogene partner 1; FGFR1 oncoprotein partner-like protein; Fgfr1op; fibroblast growth factor receptor 1 oncogene partner; FOP; LOC683722; Unknown (protein for MGC:134294)
Gene Symbols FGFR1OP
Host Species Rabbit
Immunogen Recombinant Human FGFR1OP. Recombinant protein control fragment (Product #RP-107846).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11116
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.