Learn More
Invitrogen™ FGFR1OP2 Monoclonal Antibody (1675CT713.89.84)

Description
FGFR1OP2 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | FGFR1OP2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1675CT713.89.84 |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | FGFR1OP2 |
| Gene Accession No. | Q9CRA9, Q9NVK5 |
| Gene Alias | 1500031J01Rik; FGFR1 oncogene partner 2; FGFR1 oncogene partner 2 homolog; FGFR1OP2; fibroblast growth factor receptor 1 oncogene partner 2; HSPC123; HSPC123-like; Wit3.0; Wit3.0_alpha; Wit3.0_beta; wound inducible transcript 3.0; wound inducible transcript 3.0 protein; wound-inducible transcript 3.0 protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.