Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR1OP2 Mouse anti-Human, Clone: 5C4, Bioss
SDP

Catalog No. 501979853
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9853 Supplier Bioss Supplier No. BSM51372M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May be involved in wound healing pathway.

Specifications

Antigen FGFR1OP2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5C4
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene FGFR1OP2
Gene Accession No. Q9NVK5
Gene Alias 1500031J01Rik; FGFR1 oncogene partner 2; FGFR1 oncogene partner 2 homolog; Fgfr1op2; fibroblast growth factor receptor 1 oncogene partner 2; HSPC123; HSPC123-like; Wit3.0; Wit3.0_alpha; Wit3.0_beta; wound inducible transcript 3.0; wound inducible transcript 3.0 protein; Wound-inducible transcript 3.0 protein
Gene Symbols FGFR1OP2
Host Species Mouse
Immunogen This FGFR1OP2 antibody is generated from a mouse immunized with a recombinant protein between 1-172 amino acids of human FGFR1OP2.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26127
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.