Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR2 alpha Antibody (1057960), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB111192E
Description
FGFR2 alpha Monoclonal antibody specifically detects FGFR2 alpha in Human samples. It is validated for Flow CytometrySpecifications
FGFR2 alpha | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2263 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
1057960 | |
Flow Cytometry | |
BBDS,BEK,BFR-1,CD332,CEK3,CFD1,ECT1,Fibroblast growth factor receptor,Fibroblast growth factor receptor 2,JWS,KGFR,K-SAM,Receptor protein-tyrosine kinase,TK14,TK25 | |
Chinese Hamster Ovary cell line, CHO-derived human FGFR2 alpha, Arg22-Glu377, Accession # P21802.1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction