Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR2 Antibody (016), Alexa Fluor™ 700 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289673AF700
Description
FGFR2 Monoclonal antibody specifically detects FGFR2 in Human samples. It is validated for ELISASpecifications
FGFR2 | |
Monoclonal | |
Alexa Fluor 700 | |
BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human FGFR2 extracellular domain (NP_000132.3; Met 1-Glu 377). | |
0.1 mL | |
Angiogenesis, Protein Kinase | |
2263 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
016 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction