Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR2 Antibody (7C24) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $572.50
Specifications
| Antigen | FGFR2 |
|---|---|
| Clone | 7C24 |
| Concentration | LYOPH |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018058
![]() |
Novus Biologicals
NB110941540.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB11094154
![]() |
Novus Biologicals
NB11094154 |
0.1 mg |
Each for $572.50
|
|
|||||
Description
FGFR2 Monoclonal specifically detects FGFR2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FGFR2 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
| FGFR2 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| 7C24 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rat | |
| Angiogenesis, Protein Kinase | |
| P21803 | |
| 2263 | |
| IgG2 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title