Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR2 Antibody (98739), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6843MFV610
Description
FGFR2 Monoclonal antibody specifically detects FGFR2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOFSpecifications
| FGFR2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| 98739 | |
| Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready | |
| BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms and mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms | |
| 0.1 mL | |
| Angiogenesis, Protein Kinase | |
| 2263 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction