Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR2 Antibody (OTI5C5), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270731PCP
Description
FGFR2 Monoclonal specifically detects FGFR2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
FGFR2 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
FGFR2 | |
Full length recombinant protein of human FGF R2 (NP_000132) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Flow Cytometry | |
OTI5C5 | |
Flow Cytometry | |
BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
Mouse | |
Affinity Purified | |
Angiogenesis, Protein Kinase | |
2263.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction