Learn More
Promega FGFR2 Kinase Enzyme System, 10microg
Catalog No. PRV4060 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4060 10 μg
1 options
Catalog No. PRV4060 Supplier Promega Supplier No. V4060
Recombinant human FGFR2 (285-end) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. FGFR2 is a member of the fibroblast growth factor receptor family, which play a role in mitogenesis and differentiation.
  • Easily Screen and Profile FGFR2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C