Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR2 Monoclonal Antibody (02), Invitrogen™
Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
Antigen | FGFR2 |
---|---|
Clone | 02 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543392
|
Invitrogen™
MA543392 |
50 μL |
Each for $285.50
|
|
|||||
PIMA543001
|
Invitrogen™
MA543001 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human FGFR2
FGFR2 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy. FGFR family members differ from one another in their ligand affinities and tissue distribution. Mutations in this gene are associated with Crouzon syndrome, Pfeiffer syndrome, Craniosynostosis, Apert syndrome, Jackson-Weiss syndrome, Beare-Stevenson cutis gyrata syndrome, Saethre-Chotzen syndrome, and syndromic craniosynostosis. Multiple alternatively spliced transcript variants encoding different isoforms have been noted for this gene.Specifications
FGFR2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
AU043015; AW556123; bacteria-expressed kinase; BBDS; BEK; BEK fibroblast growth factor receptor; BFR-1; CD332; CEK3; CFD1; ECT1; FGF Receptor; FGF Receptor 2; FGF-10; FGFR; FGFR2; FGFR-2; Fgfr7; Fgfr-7; fibroblast growth factor receptor 2; JWS; Keratinocyte growth factor receptor; KGFR; KGFRTr; KSAM; K-SAM; protein tyrosine kinase, receptor like 14; Soluble KGFR; svs; TK14; TK25 | |
FGFR2 | |
IgG1 | |
Protein A | |
Antibody |
02 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P21802 | |
2263 | |
Recombinant Human FGFR2 Protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
FGFR2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title