Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR2 Mouse anti-Human, Unconjugated, Clone: 1G3, Abnova™

Catalog No. 89335101 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-101 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-101 Supplier Abnova Corporation Supplier No. H00002263M01J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

FGFR2 Monoclonal antibody specifically detects FGFR2 in Human samples. It is validated for IHC-P, S-ELISA, ELISA, WB-Re, WB-Tr.

Specifications

Antigen FGFR2
Applications Immunohistochemistry (Paraffin), ELISA, Western Blot
Classification Monoclonal
Clone 1G3
Conjugate Unconjugated
Description fibroblast growth factor receptor 2
Gene FGFR2
Gene Accession No. BC039243
Gene Alias BEK, BFR-1, CD332, CEK3, CFD1, ECT1, FLJ98662, JWS, K-SAM, KGFR, TK14, TK25
Host Species Mouse
Immunogen FGFR2 (AAH39243, 621 a.a. ∼ 723 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2263
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.