Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR3 Antibody (136334), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB766MFV610
Description
FGFR3 Monoclonal antibody specifically detects FGFR3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| FGFR3 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Angiogenesis, Cancer, Protein Kinase | |
| 2261 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, CyTOF | |
| 136334 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| Pool of NS0-derived recombinant human FGF R3 alpha (IIIb) and Sf21-derived FGF R3 alpha (IIIc) | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction