Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR3 Antibody (5B6), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | FGFR3 |
---|---|
Clone | 5B6 |
Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
FGFR3 Monoclonal antibody specifically detects FGFR3 in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
FGFR3 | |
ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
A synthesized peptide derived from Human FGFR3 [UniProt P22607] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
5B6 | |
ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Protein Kinase | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
2261 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title