Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR3 Antibody (OTI1B10), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270722AF488
Description
FGFR3 Monoclonal specifically detects FGFR3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| FGFR3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FGFR3 | |
| Recombinant protein fragment corresponding to amino acids 23-375 of human FGF R3 (NP_000133) produced in SF9 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1B10 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis, Cancer, Protein Kinase | |
| 2261.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction