Learn More
Promega FGFR3 (K650E) Kinase Enzyme System, 10microg
Catalog No. PRV5082 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV5082 10 μg
1 options
Catalog No. PRV5082 Supplier Promega Supplier No. V5082
Add to Cart
Add to Cart
Recombinant human FGFR3 (K650E) (397-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Fibroblast growth factor receptor 3 is part of a family of fibroblast growth factor receptors that share similar structure and function
  • Easily Screen and Profile FGFR3 (K650E) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C