Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR3 Mouse anti-Human, Clone:1E3F9, Abnova™

Catalog No. 89333096 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-096 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-096 Supplier Abnova Corporation Supplier No. MAB16731
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human FGFR3.

This gene encodes a member of the fibroblast growth factor receptor (FGFR) family, with its amino acid sequence being highly conserved between members and among divergent species. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein would consist of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. This particular family member binds acidic and basic fibroblast growth hormone and plays a role in bone development and maintenance. Mutations in this gene lead to craniosynostosis and multiple types of skeletal dysplasia. Three alternatively spliced transcript variants that encode different protein isoforms have been described. [provided by RefSeq]

Specifications

Antigen FGFR3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E3F9
Conjugate Unconjugated
Description fibroblast growth factor receptor 3
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene FGFR3
Gene Alias ACH, CD333, CEK2, HSFGFR3EX, JTK4
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 529-694 of human FGFR3 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 2261
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.