Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR3 Rabbit anti-Mouse, Clone: 42, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290392100UL
Description
FGFR3 Monoclonal specifically detects FGFR3 in Mouse samples. It is validated for Sandwich ELISA Capture.Specifications
| FGFR3 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 42 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| Recombinant Mouse FGFR3 Protein | |
| 100 μg | |
| Angiogenesis, Cancer, Protein Kinase | |
| 2261 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction