Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR4 Antibody (148), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290949JF646
Description
FGFR4 Monoclonal antibody specifically detects FGFR4 in Macaque samples. It is validated for ELISASpecifications
FGFR4 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Macaque | |
Purified |
ELISA | |
148 | |
ELISA | |
CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rhesus FGFR4 (Accession#: XP_001087243.1; Met1-Asp369). | |
0.1 mL | |
Angiogenesis, Protein Kinase | |
2264 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction