Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR4 Antibody (240929) [Biotin], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6852B
Description
FGFR4 Monoclonal antibody specifically detects FGFR4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOFSpecifications
FGFR4 | |
Monoclonal | |
Biotin | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
240929 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready | |
CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase | |
Mouse myeloma cell line NS0-derived recombinant human FGF R4, Leu22-Asp369 (predicted), Accession # P22455 | |
0.1 mL | |
Angiogenesis, Protein Kinase | |
2264 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction