Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR4 Antibody (240929) [PerCP], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6852C
Description
FGFR4 Monoclonal antibody specifically detects FGFR4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOFSpecifications
| FGFR4 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 240929 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready | |
| CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase | |
| Mouse myeloma cell line NS0-derived recombinant human FGF R4, Leu22-Asp369 (predicted), Accession # P22455 | |
| 0.1 mL | |
| Angiogenesis, Protein Kinase | |
| 2264 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction