Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR4 Antibody (8A5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | FGFR4 |
---|---|
Clone | 8A5 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
FGFR4 Monoclonal antibody specifically detects FGFR4 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
FGFR4 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase | |
A synthesized peptide derived from Human FGFR4 [UniProt P22455] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
8A5 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Angiogenesis, Protein Kinase | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
2264 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title