Learn More
Promega FGFR4 Kinase Enzyme System, 1mg
Catalog No. PRV6393 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6393 1 mg
1 options
Catalog No. PRV6393 Supplier Promega Supplier No. V6393
Add to Cart
Add to Cart
Request a Quote
Recombinant human FGFR4 (460-end) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. FGFR4 is a member of the fibroblast growth factor receptor family, which play a role in mitogenesis and differentiation.
  • Easily Screen and Profile FGFR4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C