Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR5/FGFRL1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298227PCP
Description
FGFR5/FGFRL1 Polyclonal antibody specifically detects FGFR5/FGFRL1 in Mouse samples. It is validated for ELISASpecifications
FGFR5/FGFRL1 | |
Polyclonal | |
ELISA | |
FGF homologous factor receptor, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1 | |
Produced in rabbits immunized with purified, recombinant Mouse FGFR5/FGFRL1 (Accession#: NP_473412.1; Met 1-Pro 374) | |
0.1 mL | |
Angiogenesis | |
53834 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction