Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR5/FGFRL1 Antibody (RM0232-7L43) - Azide and BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $560.50

Specifications

Antigen FGFR5/FGFRL1
Clone RM0232-7L43
Concentration LYOPH
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019149
SDP
View Documents
Novus Biologicals
NBP2122940.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP212294
SDP
View Documents
Novus Biologicals
NBP212294
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

FGFR5/FGFRL1 Monoclonal specifically detects FGFR5/FGFRL1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

FGFR5/FGFRL1
LYOPH
Monoclonal
Purified
RUO
Mouse
FGF homologous factor receptor, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1
FGFRL1
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
RM0232-7L43
Western Blot
Unconjugated
Rat
Angiogenesis
Q91V87
53834
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.