Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGFR5/FGFRL1 Antibody (RM0232-7L43), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212294B
Description
FGFR5/FGFRL1 Monoclonal specifically detects FGFR5/FGFRL1 in Mouse samples. It is validated for Western Blot.Specifications
FGFR5/FGFRL1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
FGFRL1 | |
Mouse recombinant FGFR5/FGFRL1 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot | |
RM0232-7L43 | |
Western Blot | |
FGF homologous factor receptor, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1 | |
Rat | |
Protein A or G purified | |
Angiogenesis | |
53834 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction