Learn More
FGFR5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA547218
Description
In Western blots, less than 5% cross-reactivity with recombinant human (rh) FGF R1 alpha (IIIb), recombinant mouse (rm) FGF R2 beta (IIIc), rmFGF R3 alpha (IIIb), rhFGF R3 alpha (IIIc), rhFGF R1 beta (IIIc), rmFGF R2 beta (IIIb), rhFGF R1 alpha (IIIc), rmFGF R2 alpha (IIIc), rhFGF R1 beta (IIIb), rmFGF R2 alpha (IIIb), rmFGF R2, and rmFGF R4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
FGFR5 Polyclonal antibody specifically detects FGFR5 in Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.
Specifications
FGFR5 | |
Polyclonal | |
Unconjugated | |
FGFRL1 | |
alpha-L-iduronidase; FGF homologous factor receptor; FGF receptor-like protein 1; Fgfr5; FGFR-5; FGFR5beta; FGFR5gamma; Fgfrl1; FGFR-like protein; FHFR; fibroblast growth factor receptor 5; Fibroblast growth factor receptor-like 1; fibroblast growth factor receptor-like protein; UNQ480/PRO943 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
116701 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q91V87 | |
FGFRL1 | |
Mouse myeloma cell line NS0-derived recombinant mouse FGF R5 beta Ala21-Pro374 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.