Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fggy Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120316 Supplier Invitrogen™ Supplier No. PA5120316
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, HepG2, A-431, Mouse kidney, Rat liver. Immunogen sequence: MSGGEQKPER YYVGVDVGTG SVRAALVDQS GVLLAFADQP IKNWEPQFNH HEQSSEDIWA ACCVVTKKVV QGIDLNQIRG LGFDATCSLV VLDKQFHPLP VNQEGDSHRN VIMWLDHRAV SQVNRINETK HSVLQYVGGV MSVEMQAPKL LWLKENLREI CWDKAGHFFD LPDFLSWKAT GVTARSLCSL VCKWTYSAEK GWDDSFWKMI GLEDFVADNY SKIGNQVLPP GASLGNGLTP EAARDLGLLP.

FGGY gene ontology annotations related to this gene include neuron cellular homeostasis; pentose metabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fggy
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FGGY
Gene Accession No. A2AJL3, Q96C11
Gene Alias 2310009E04Rik; Fggy; FGGY carbohydrate kinase domain containing; FGGY carbohydrate kinase domain-containing protein; zgc:85818
Gene Symbols FGGY
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human FGGY (NP_060761.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298250, 55277, 75578
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.