Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FGL1 Monoclonal Antibody (C7), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $458.50
Specifications
Antigen | FGL1 |
---|---|
Clone | C7 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA551084
|
Invitrogen™
MA551084 |
20 μL |
Each for $195.80
|
|
|||||
FEMA551083
|
Invitrogen™
MA551083 |
200 μL |
Each for $458.50
|
|
|||||
Description
Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen beta- and gamma- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas. Four alternatively spliced transcript variants encoding the same protein exist for this gene.Specifications
FGL1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
Fgl1; fibrinogen like 1; fibrinogen-like 1; fibrinogen-like protein 1; Fibrinogen-related protein 1; fibronigen-like protein 1; Frep1; Hepassocin; hepatocellular carcinoma-related sequence; hepatocyte-derived fibrinogen-related protein 1; HFREP1; HFREP-1; HP-041; HPS; Lfire1; LFIRE-1; Liver fibrinogen-related protein 1; Liver fibrinogen-related protein-1; Mfire1; Mfire-1; Mfrep-1; MGC12455; OTTHUMP00000122468 | |
FGL1 | |
IgG1 κ | |
Protein A/G | |
FGL1 |
C7 | |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q08830 | |
2267 | |
Recombinant Human FGL1 protein, Leu23-Ile312 (Accession #Q08830). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title