Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547068
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547068 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547068 Supplier Invitrogen™ Supplier No. PA547068
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen beta- and gamma- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas. Four alternatively spliced transcript variants encoding the same protein exist for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGL1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene FGL1
Gene Accession No. Q5M8C6
Gene Alias Fgl1; fibrinogen like 1; fibrinogen-like 1; fibrinogen-like protein 1; Fibrinogen-related protein 1; fibronigen-like protein 1; Frep1; Hepassocin; hepatocellular carcinoma-related sequence; hepatocyte-derived fibrinogen-related protein 1; HFREP1; HFREP-1; HP-041; HPS; Lfire1; LFIRE-1; Liver fibrinogen-related protein 1; Liver fibrinogen-related protein-1; Mfire1; Mfire-1; Mfrep-1; MGC12455; OTTHUMP00000122468
Gene Symbols FGL1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Hepassocin Asp25-Val314.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246186
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.