Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FGL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | FGL1 |
---|---|
Concentration | 0.71 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119116
|
Invitrogen™
PA5119116 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121993
|
Invitrogen™
PA5121993 |
20 μL |
Each for $209.50
|
|
|||||
Description
FGL1 Polyclonal Antibody for Western Blot, IHC (P)
Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen beta- and gamma- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas. Four alternatively spliced transcript variants encoding the same protein exist for this gene.Specifications
FGL1 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
FGL1 | |
Fgl1; fibrinogen like 1; fibrinogen-like 1; fibrinogen-like protein 1; Fibrinogen-related protein 1; fibronigen-like protein 1; Frep1; Hepassocin; hepatocellular carcinoma-related sequence; hepatocyte-derived fibrinogen-related protein 1; HFREP1; HFREP-1; HP-041; HPS; Lfire1; LFIRE-1; Liver fibrinogen-related protein 1; Liver fibrinogen-related protein-1; Mfire1; Mfire-1; Mfrep-1; MGC12455; OTTHUMP00000122468 | |
FGL1 | |
Primary | |
Antigen Affinity Chromatography |
0.71 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q08830 | |
2267 | |
Recombinant protein Fibrinogen Like Protein 1. The antigen corresponds to amino acid range 23-312 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title