Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555376 Supplier Invitrogen™ Supplier No. PA555376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQEQIQSRPV QHLIYKDCSD YYAIGKRSSE TYRVTPDPKN SSFEVYCDME TMGGGWTVLQ ARLDGSTNFT RTWQDYKAGF GNLRR Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

Fibrinogen-like protein 2 (Fgl2), also known as fibroleukin, is secreted by T cells and is involved in diseases in which thrombosis plays a pivotal role, such as virus-induced fulminant hepatitis, fetal loss syndrome and xenograft rejection. Constitutively expressed in cytotoxic T cells, Fgl2 exerts immunosuppressive effects on both T cell proliferation and dendritic cell maturation. Fgl2 is a serine protease and directly cleaves prothrombin to thrombin. Fgl2 functions in the pathogenesis of diseases including viral-induced hepatitis and Th1 cytokine-induced fetal loss syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FGL2
Gene Accession No. Q14314
Gene Alias AI385601; associated with MHV-3 induced prothrombinase activity; cytotoxic T-lymphocyte-specific protein; Fgl2; Fiblp; fibrinogen like 2; fibrinogen-like 2; fibrinogen-like protein 2; Fibroleukin; musfiblp; Prothrombinase; pT49; similar to Fibroleukin precursor (Fibrinogen-like protein 2) (pT49); T49
Gene Symbols FGL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FGL2. Recombinant protein control fragment (Product #RP-92705).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10875
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.