Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FGL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571472
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571472 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571472 Supplier Invitrogen™ Supplier No. PA571472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FGL2 Polyclonal Antibody for Flow, Western Blot

Fibrinogen-like protein 2 (Fgl2), also known as fibroleukin, is secreted by T cells and is involved in diseases in which thrombosis plays a pivotal role, such as virus-induced fulminant hepatitis, fetal loss syndrome and xenograft rejection. Constitutively expressed in cytotoxic T cells, Fgl2 exerts immunosuppressive effects on both T cell proliferation and dendritic cell maturation. Fgl2 is a serine protease and directly cleaves prothrombin to thrombin. Fgl2 functions in the pathogenesis of diseases including viral-induced hepatitis and Th1 cytokine-induced fetal loss syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGL2
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FGL2
Gene Accession No. P12804, Q14314
Gene Alias AI385601; associated with MHV-3 induced prothrombinase activity; cytotoxic T-lymphocyte-specific protein; Fgl2; Fiblp; fibrinogen like 2; fibrinogen-like 2; fibrinogen-like protein 2; Fibroleukin; musfiblp; Prothrombinase; pT49; similar to Fibroleukin precursor (Fibrinogen-like protein 2) (pT49); T49
Gene Symbols FGL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 338-367 amino acids from the C-terminal region of human FGL2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10875, 14190
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.