Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FH 1

Catalog No. 525410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-541-0 10 mg
52-545-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-541-0 Supplier Tocris Bioscience™ Supplier No. 5254/10
This item is not returnable. View return policy

Enhances iPSC-derived hepatocyte differentiation and maturation

FH 1 enhances differentiation of induced pluripotent stem cells (iPSCs) to hepatocytes; also promotes the maturation of iPSC-derived hepatocytes.

Specifications

Purity >99%
Quantity 10 mg
Product Type FH 1
Molecular Weight (g/mol) 282.34
Inhibitors FH 1

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.