Learn More
Description
Specifications
Specifications
| Antigen | fHbp |
| Applications | ELISA, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | JAR 4 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal antibody IgG2a in PBS without azide |
| Host Species | Mouse |
| Immunogen | His-tagged recombinant Factor H-binding protein (fHbp) from variant group 1 expressed in E. coli. |
| Purification Method | Protein G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.