Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

fHbp Mouse anti-Bacteria, Clone: JAR 4, MilliporeSigma™

Catalog No. MABF267025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF267025 25 μg
MABF2670100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF267025 Supplier MilliporeSigma Supplier No. MABF267025UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

fHbp Monoclonal antibody specifically detects fHbp in Bacteria samples. It is validated for Activity Assay, ELISA, Flow Cytometry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen fHbp
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone JAR 4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in PBS without azide
Host Species Mouse
Immunogen His-tagged recombinant Factor H-binding protein (fHbp) from variant group 1 expressed in E. coli.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.