Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

fHbp Mouse anti-Bacteria, Clone: JAR 5, MilliporeSigma™

Catalog No. MABF266925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF266925 25 μL
MABF2669100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF266925 Supplier MilliporeSigma Supplier No. MABF266925UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

fHbp Monoclonal antibody specifically detects fHbp in Bacteria samples. It is validated for Dot Blot, ELISA, Inhibition, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen fHbp
Applications Dot Blot, ELISA, Inhibition Assays, Western Blot
Classification Monoclonal
Clone JAR 5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in PBS without azide.
Host Species Mouse
Immunogen His-tagged recombinant Factor H-binding protein (fHbp) from variant group 1, strain MC58, expressed in E. coli.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.