Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FHIT Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300295PCP
Description
FHIT Polyclonal antibody specifically detects FHIT in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
FHIT | |
Polyclonal | |
ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
AP3A hydrolase, AP3AaseFRA3Bbis(5'-adenosyl)-triphosphatase, Diadenosine 5'-5'''-P1, dinucleosidetriphosphatase, EC 3.6.1.29, fragile histidine triad gene, Fragile histidine triad protein, P3-triphosphate hydrolase, tumor suppressor protein | |
Produced in rabbits immunized with purified, recombinant Human FHIT (Accession#: P49789; Met1-Gln147) | |
0.1 mL | |
Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Tumor Suppressors | |
2272 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction