Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FHIT Antibody (SR1259), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | FHIT |
---|---|
Clone | SR1259 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
FHIT Monoclonal antibody specifically detects FHIT in Human, Rat samples. It is validated for Western BlotSpecifications
FHIT | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
AP3A hydrolase, AP3AaseFRA3Bbis(5'-adenosyl)-triphosphatase, Diadenosine 5'-5'''-P1, dinucleosidetriphosphatase, EC 3.6.1.29, fragile histidine triad gene, Fragile histidine triad protein, P3-triphosphate hydrolase, tumor suppressor protein | |
A synthesized peptide derived from human FHIT (Uniprot #: P49789) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1259 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2272 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title